Assay Details
Gene Symbol: BEND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184767.1 | NP_001171696.1 | 8 - 9 | 1506 | 123 | |
NM_153346.4 | NP_699177.2 | 10 - 11 | 1779 | 123 | ||
XM_011545447.1 | XP_011543749.1 | 10 - 11 | 1803 | 123 | ||
XM_017029266.1 | XP_016884755.1 | 9 - 10 | 1677 | 123 | ||
XM_017029267.1 | XP_016884756.1 | 10 - 11 | 1803 | 123 | ||
GenBank mRNA | AK128155.1 | - | 10 - 11 | 1761 | 123 | |
BC037301.2 | - | 10 - 11 | 1759 | 4827886 | 123 | |
BC096745.1 | - | 8 - 9 | 1439 | 4830576 | 123 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000893.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X011939.hg.1 | TC0X000893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011939.hg.1 | ENST00000380030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011939.hg.1 | ENST00000380033.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011939.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011939.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162950.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162950.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162950.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162950.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162950.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162950.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162950.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162950.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162950.hg.1 | TC0X00009184.hg.1 | Clariom D TC |
