Assay Details
Gene Symbol: BEND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153346.4 | NP_699177.2 | 13 - 14 | 2342 | 124 | |
XM_011545447.1 | XP_011543749.1 | 13 - 14 | 2366 | 124 | ||
XM_017029266.1 | XP_016884755.1 | 12 - 13 | 2240 | 124 | ||
GenBank mRNA | AK128155.1 | - | 13 - 14 | 2324 | 124 | |
BC037301.2 | - | 13 - 14 | 2322 | 4827886 | 124 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000893.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011932.hg.1 | TC0X000893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011932.hg.1 | ENST00000380033.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011932.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011934.hg.1 | TC0X000893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011934.hg.1 | ENST00000380033.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011934.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011930.hg.1 | TC0X000893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011930.hg.1 | ENST00000380033.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011930.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162944.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162944.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162944.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162944.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162944.hg.1 | TC0X00009184.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162945.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162945.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162945.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162945.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162945.hg.1 | TC0X00009184.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 227904 | - | - |
