Assay Details
Gene Symbol: BEND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153346.4 | NP_699177.2 | 7 - 8 | 1340 | 113 | |
GenBank mRNA | AK128155.1 | - | 7 - 8 | 1322 | 113 | |
BC037301.2 | - | 7 - 8 | 1320 | 4827886 | 113 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000893.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X011942.hg.1 | TC0X000893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011942.hg.1 | ENST00000380030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011942.hg.1 | ENST00000380033.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011942.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011942.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162953.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162953.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162953.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162953.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162953.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162953.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162953.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162953.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162953.hg.1 | TC0X00009184.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380030.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | ENST00000380033.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_001184767.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009184.hg.1 | NM_153346.4 | Refseq |
