| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
ENST00000366667.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
ENST00000366667.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
ENST00000366667.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
ENST00000366667.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
ENST00000366667.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
ENST00000366667.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
ENST00000366667.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
ENST00000366667.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
NM_000029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
NM_000029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
NM_000029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
NM_000029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
NM_000029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
NM_000029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
NM_000029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
NM_000029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018565.hg.1 |
NM_000029.3 |
Refseq |