Assay Details
| Gene Symbol: TSPAN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290212.1 | NP_001277141.1 | 1 - 2 | 238 | 196 | |
| GenBank mRNA | AB593081.1 | - | 1 - 2 | 211 | 196 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB593081.1,NM_001290212.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169580.hg.1 | TC1700009184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169573.hg.1 | ENST00000571914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169573.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169573.hg.1 | ENST00000571914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169573.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169573.hg.1 | TC1700009184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | ENST00000611590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | ENST00000611590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169581.hg.1 | TC1700009184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169574.hg.1 | ENST00000571914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169574.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169574.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169574.hg.1 | ENST00000571914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169574.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169574.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169574.hg.1 | TC1700009184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | ENST00000611590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | ENST00000571707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | ENST00000574882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | ENST00000611590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | ENST00000621293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | NM_001290212.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | NM_031945.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | NR_110866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169582.hg.1 | TC1700009184.hg.1 | Clariom D TC |

TSPAN10
Genomic Map