Assay Details
| Gene Symbol: ARNTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030272.2 | NP_001025443.1 | 13 - 14 | 1579 | 155 | |
| NM_001030273.2 | NP_001025444.1 | 14 - 15 | 1650 | 155 | ||
| NM_001178.5 | NP_001169.3 | 14 - 15 | 1634 | 155 | ||
| NM_001297719.1 | NP_001284648.1 | 14 - 15 | 1637 | 155 | ||
| NM_001297722.1 | NP_001284651.1 | 13 - 14 | 1582 | 155 | ||
| NM_001297724.1 | NP_001284653.1 | 14 - 15 | 1646 | 155 | ||
| XM_006718233.3 | XP_006718296.1 | 13 - 14 | 1524 | 155 | ||
| XM_006718234.1 | XP_006718297.1 | 14 - 15 | 1590 | 155 | ||
| XM_011520105.2 | XP_011518407.2 | 12 - 13 | 1685 | 155 | ||
| XM_011520107.2 | XP_011518409.2 | 12 - 13 | 1680 | 155 | ||
| XM_011520108.1 | XP_011518410.1 | 14 - 15 | 1630 | 155 | ||
| XM_011520109.1 | XP_011518411.1 | 12 - 13 | 1385 | 155 | ||
| XM_011520110.1 | XP_011518412.1 | 15 - 16 | 1645 | 155 | ||
| XM_011520111.1 | XP_011518413.1 | 13 - 14 | 1460 | 155 | ||
| XM_011520112.2 | XP_011518414.1 | 15 - 16 | 1645 | 155 | ||
| XM_011520113.1 | XP_011518415.1 | 15 - 16 | 1642 | 155 | ||
| XM_017017738.1 | XP_016873227.1 | 12 - 13 | 1686 | 155 | ||
| XM_017017739.1 | XP_016873228.1 | 12 - 13 | 1682 | 155 | ||
| XM_017017740.1 | XP_016873229.1 | 13 - 14 | 1521 | 155 | ||
| XM_017017741.1 | XP_016873230.1 | 12 - 13 | 1382 | 155 | ||
| XM_017017742.1 | XP_016873231.1 | 12 - 13 | 1385 | 155 | ||
| XM_017017743.1 | XP_016873232.1 | 12 - 13 | 1382 | 155 | ||
| XM_017017744.1 | XP_016873233.1 | 15 - 16 | 1642 | 155 | ||
| XM_017017745.1 | XP_016873234.1 | 14 - 15 | 1587 | 155 | ||
| XM_017017746.1 | XP_016873235.1 | 14 - 15 | 1600 | 155 | ||
| XM_017017747.1 | XP_016873236.1 | 13 - 14 | 1460 | 155 | ||
| XM_017017748.1 | XP_016873237.1 | 13 - 14 | 1514 | 155 | ||
| GenBank mRNA | AB000812.1 | - | 11 - 12 | 1218 | 155 | |
| AB000815.1 | - | 12 - 13 | 1071 | 155 | ||
| AF044288.1 | - | 14 - 15 | 1547 | 155 | ||
| AK095749.1 | - | 14 - 15 | 1535 | 155 | ||
| AK291510.1 | - | 13 - 14 | 1495 | 155 | ||
| AK315174.1 | - | 14 - 15 | 1575 | 155 | ||
| BC016674.1 | - | 11 - 12 | 1181 | 3921870 | 155 | |
| BC031214.1 | - | 14 - 15 | 1478 | 4793007 | 155 | |
| BC041129.1 | - | 14 - 15 | 1558 | 5753529 | 155 | |
| D89722.1 | - | 11 - 12 | 1194 | 155 | ||
| U51627.1 | - | 8 - 9 | 1176 | 155 | ||
| U60415.1 | - | 13 - 14 | 1481 | 155 | ||
More Information
Set Membership: |
|

ARNTL
Genomic Map