Assay Details
Gene Symbol: ARNTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030272.2 | NP_001025443.1 | 15 - 16 | 1814 | 149 | |
NM_001030273.2 | NP_001025444.1 | 16 - 17 | 1885 | 149 | ||
NM_001178.5 | NP_001169.3 | 16 - 17 | 1869 | 149 | ||
NM_001297719.1 | NP_001284648.1 | 16 - 17 | 1872 | 149 | ||
NM_001297722.1 | NP_001284651.1 | 15 - 16 | 1817 | 149 | ||
NM_001297724.1 | NP_001284653.1 | 16 - 17 | 1881 | 149 | ||
XM_011520107.2 | XP_011518409.2 | 14 - 15 | 1915 | 149 | ||
XM_011520112.2 | XP_011518414.1 | 17 - 18 | 1880 | 149 | ||
XM_011520113.1 | XP_011518415.1 | 17 - 18 | 1877 | 149 | ||
XM_017017739.1 | XP_016873228.1 | 14 - 15 | 1917 | 149 | ||
XM_017017742.1 | XP_016873231.1 | 14 - 15 | 1620 | 149 | ||
XM_017017743.1 | XP_016873232.1 | 14 - 15 | 1617 | 149 | ||
XM_017017746.1 | XP_016873235.1 | 16 - 17 | 1835 | 149 | ||
XM_017017747.1 | XP_016873236.1 | 15 - 16 | 1695 | 149 | ||
XM_017017748.1 | XP_016873237.1 | 15 - 16 | 1749 | 149 | ||
GenBank mRNA | AB000812.1 | - | 13 - 14 | 1453 | 149 | |
AB000814.1 | - | 2 - 3 | 269 | 149 | ||
AF044288.1 | - | 16 - 17 | 1782 | 149 | ||
AK095749.1 | - | 16 - 17 | 1770 | 149 | ||
AK128317.1 | - | 2 - 3 | 172 | 149 | ||
AK291510.1 | - | 15 - 16 | 1730 | 149 | ||
AK315174.1 | - | 16 - 17 | 1810 | 149 | ||
BC016674.1 | - | 13 - 14 | 1416 | 3921870 | 149 | |
BC031214.1 | - | 16 - 17 | 1713 | 4793007 | 149 | |
BC041129.1 | - | 16 - 17 | 1793 | 5753529 | 149 | |
D89722.1 | - | 13 - 14 | 1429 | 149 | ||
U51627.1 | - | 10 - 11 | 1411 | 149 | ||
U60415.1 | - | 15 - 16 | 1716 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
