Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019705.hg.1 |
TC06001663.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019705.hg.1 |
ENST00000338863.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019705.hg.1 |
ENST00000373905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019705.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000402886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
ENST00000433586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011631.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179691.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179691.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179691.hg.1 |
ENST00000338863.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179691.hg.1 |
NM_003214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179691.hg.1 |
TC0600011631.hg.1 |
Clariom D TC |