Assay Details
Gene Symbol: TEAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003214.3 | NP_003205.2 | 9 - 10 | 964 | 136 | |
GenBank mRNA | AF142482.1 | - | 9 - 10 | 918 | 136 | |
AK226144.1 | - | 5 - 6 | 2324 | 136 | ||
BC010053.2 | - | 9 - 10 | 820 | 136 | ||
BC027877.1 | - | 7 - 8 | 702 | 5207501 | 136 | |
BC091488.1 | - | 9 - 10 | 889 | 6187474 | 136 | |
X94439.1 | - | 8 - 9 | 728 | 136 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001663.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
female pregnancy
hippo signaling
positive regulation of transcription from RNA polymerase II promoter
asymmetric neuroblast division
RNA polymerase II transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
female pregnancy
hippo signaling
positive regulation of transcription from RNA polymerase II promoter
asymmetric neuroblast division
RNA polymerase II transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06019694.hg.1 | TC06001663.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06019694.hg.1 | ENST00000338863.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06019694.hg.1 | ENST00000373905.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06019694.hg.1 | ENST00000402886.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06019694.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06019695.hg.1 | TC06001663.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06019695.hg.1 | ENST00000338863.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06019695.hg.1 | ENST00000373905.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06019695.hg.1 | ENST00000402886.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06019695.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06019693.hg.1 | TC06001663.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06019693.hg.1 | ENST00000338863.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06019693.hg.1 | ENST00000373905.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06019693.hg.1 | ENST00000402886.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06019693.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179685.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179685.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179685.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179685.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179685.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179685.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179685.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179685.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179685.hg.1 | TC0600011631.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011631.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179684.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179684.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179684.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179684.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179684.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179684.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179684.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179684.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179684.hg.1 | TC0600011631.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600179683.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179683.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179683.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179683.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179683.hg.1 | ENST00000338863.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179683.hg.1 | ENST00000402886.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179683.hg.1 | ENST00000433586.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179683.hg.1 | NM_003214.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179683.hg.1 | TC0600011631.hg.1 | Clariom D TC |
