Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017924.hg.1 |
ENST00000291440.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017924.hg.1 |
NM_015692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017924.hg.1 |
TC19001268.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900181603.hg.1 |
ENST00000388925.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181603.hg.1 |
ENST00000443236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181603.hg.1 |
NM_015692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181603.hg.1 |
TC1900009941.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900181603.hg.1 |
ENST00000388925.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181603.hg.1 |
ENST00000443236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181603.hg.1 |
NM_015692.3 |
Refseq |