Assay Details
| Gene Symbol: FLJ25758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024372.1 | 1 | 1276 | 143 | ||
| GenBank mRNA | AK098624.1 | - | 1 | 1283 | 143 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
adhesion G protein-coupled receptor E4, pseudogene |
ADGRE4P |
EMR4, EMR4P, FIRE, GPR127, PGR16 |
-14.1kb | Sense | Chr. 19 - 6952500 - 6990846 on NCBI Build GRCh38 |
| Downstream | methyl-CpG binding domain protein 3 like 5 | MBD3L5 | 23.5kb | Anti-sense | Chr. 19 - 7030583 - 7033011 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FLJ25758
Genomic Map