Assay Details
Gene Symbol: CERS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203463.2 | NP_982288.1 | 9 - 10 | 1202 | 122 | |
GenBank mRNA | AK172775.1 | - | 9 - 10 | 1202 | 122 | |
BC030800.2 | - | 4 - 5 | 433 | 122 | ||
BG206657.1 | - | 6 - 7 | 608 | 122 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000996.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC030800.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02014546.hg.1 | TC02000996.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02014546.hg.1 | ENST00000305747.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014546.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014546.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014546.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014548.hg.1 | TC02000996.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02014548.hg.1 | ENST00000305747.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014548.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014548.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014548.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014543.hg.1 | TC02000996.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02014543.hg.1 | ENST00000305747.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014543.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014543.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014543.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014544.hg.1 | TC02000996.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02014544.hg.1 | ENST00000305747.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014544.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014544.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014544.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014545.hg.1 | TC02000996.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02014545.hg.1 | ENST00000305747.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014545.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014545.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014545.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014547.hg.1 | TC02000996.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02014547.hg.1 | ENST00000305747.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014547.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014547.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014547.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014549.hg.1 | TC02000996.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02014549.hg.1 | ENST00000305747.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014549.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014549.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014549.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169079.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169079.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169079.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169079.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169079.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169079.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169079.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169079.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169079.hg.1 | TC0200009895.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200169081.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169081.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169081.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169081.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169081.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169081.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169081.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169081.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169081.hg.1 | TC0200009895.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169078.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169078.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169078.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169078.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169078.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169078.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169078.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169078.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169078.hg.1 | TC0200009895.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200169080.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169080.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169080.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169080.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169080.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169080.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169080.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169080.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169080.hg.1 | TC0200009895.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200169077.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169077.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169077.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169077.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169077.hg.1 | ENST00000305747.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169077.hg.1 | ENST00000392687.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169077.hg.1 | NM_001256126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169077.hg.1 | NM_203463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169077.hg.1 | TC0200009895.hg.1 | Clariom D TC |
