Assay Details
Gene Symbol: CYCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018947.5 | NP_061820.1 | 1 - 2 | 152 | 152 | |
GenBank mRNA | BU597562.1 | - | 1 - 2 | 130 | 152 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001207.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_018947.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
mitochondrial electron transport, ubiquinol to cytochrome c
mitochondrial electron transport, cytochrome c to oxygen
protein dephosphorylation
mitochondrion organization
activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
cellular respiration
intrinsic apoptotic signaling pathway
protein serine/threonine phosphatase activity
protein binding
heme binding
electron transporter, transferring electrons from CoQH2-cytochrome c reductase complex and cytochrome c oxidase complex activity
metal ion binding
response to reactive oxygen species
mitochondrial electron transport, ubiquinol to cytochrome c
mitochondrial electron transport, cytochrome c to oxygen
protein dephosphorylation
mitochondrion organization
activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
cellular respiration
intrinsic apoptotic signaling pathway
protein serine/threonine phosphatase activity
protein binding
heme binding
electron transporter, transferring electrons from CoQH2-cytochrome c reductase complex and cytochrome c oxidase complex activity
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00008800.hg.1 | ENST00000402289.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018903.hg.1 | TC07001207.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07018903.hg.1 | ENST00000305786.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018903.hg.1 | ENST00000409409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018903.hg.1 | ENST00000409764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018903.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007746.hg.1 | ENST00000399027.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007746.hg.1 | ENST00000399027.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006654.hg.1 | ENST00000427212.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006654.hg.1 | ENST00000427212.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013640.hg.1 | ENST00000595083.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006988.hg.1 | ENST00000558168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010449.hg.1 | ENST00000525280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010405.hg.1 | ENST00000517555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009946.hg.1 | ENST00000438233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018908.hg.1 | TC07001207.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07018908.hg.1 | ENST00000305786.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018908.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010091.hg.1 | ENST00000523393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009927.hg.1 | ENST00000443545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008932.hg.1 | ENST00000411715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007699.hg.1 | ENST00000534107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008637.hg.1 | ENST00000448560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018899.hg.1 | TC07001207.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07018899.hg.1 | ENST00000305786.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018899.hg.1 | ENST00000409409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018899.hg.1 | ENST00000409764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018899.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006848.hg.1 | ENST00000420810.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008859.hg.1 | ENST00000407870.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174744.hg.1 | TC0700010509.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009927.hg.1 | ENST00000443545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013640.hg.1 | ENST00000595083.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010509.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010091.hg.1 | ENST00000523393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010405.hg.1 | ENST00000517555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006654.hg.1 | ENST00000427212.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006654.hg.1 | ENST00000427212.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007746.hg.1 | ENST00000399027.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007746.hg.1 | ENST00000399027.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006848.hg.1 | ENST00000420810.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010449.hg.1 | ENST00000525280.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006988.hg.1 | ENST00000558168.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | ENST00000409409.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | ENST00000409764.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | ENST00000413447.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174749.hg.1 | TC0700010509.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00009946.hg.1 | ENST00000438233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007699.hg.1 | ENST00000534107.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174754.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174754.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174754.hg.1 | ENST00000305786.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174754.hg.1 | NM_018947.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174754.hg.1 | TC0700010509.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600008859.hg.1 | ENST00000407870.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008932.hg.1 | ENST00000411715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008800.hg.1 | ENST00000402289.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008637.hg.1 | ENST00000448560.1 | Ensembl |
