Assay Details
| Gene Symbol: LOC101929066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125429.1 | 3 - 4 | 373 | 95 | ||
| NR_125430.1 | 2 - 3 | 129 | 95 | |||
| GenBank mRNA | BX402012.2 | - | 3 - 4 | 373 | 95 | |
| CA396779.1 | - | 2 - 3 | 496 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006885.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146685.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146685.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146685.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146685.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146685.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146685.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146685.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146685.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146685.hg.1 | TC0800006885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146683.hg.1 | TC0800006885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800146686.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146686.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146686.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146686.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146686.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146686.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146686.hg.1 | TC0800006885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000499554.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000505114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000517798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | ENST00000521775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006885.hg.1 | NR_125430.1 | Refseq |

LOC101929066
Genomic Map