Assay Details
| Gene Symbol: GRIK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027021.1 | 3 - 4 | 613 | 137 | ||
| GenBank mRNA | BG187681.1 | - | 3 - 4 | 618 | 137 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GRIK1 antisense RNA 2 | GRIK1-AS2 |
C21orf41, NCRNA00258 |
-128.1kb | Sense | Chr. 21 - 29596040 - 29630747 on NCBI Build GRCh38 |
| Downstream | claudin 17 | CLDN17 | 405.9kb | Anti-sense | Chr. 21 - 30165923 - 30166653 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000100.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000636.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000636.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000636.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000636.hg.1 | TC21000100.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | TC2100006869.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146283.hg.1 | NR_027021.1 | Refseq |

GRIK1-AS1
Genomic Map