Assay Details
| Gene Symbol: TNNC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011529031.2 | XP_011527333.1 | 2 - 3 | 130 | 152 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011529031.2 transcripts.
Set Membership: |
|

TNNC2
Genomic Map