Assay Details
| Gene Symbol: RBM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017016103.1 | XP_016871592.1 | 1 | 1079 | 56 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | structural maintenance of chromosomes 3 | SMC3 |
BAM, BMH, CDLS3, CSPG6, HCAP, SMC3L1 |
-37.3kb | Sense | Chr. 10 - 110567691 - 110604634 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017016103.1 transcripts.
Set Membership: |
|

RBM20
Genomic Map