Assay Details
| Gene Symbol: NEK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199397.1 | NP_001186326.1 | 18 - 19 | 2142 | 98 | |
| NM_012224.2 | NP_036356.1 | 17 - 18 | 2220 | 98 | ||
| XM_006714228.1 | XP_006714291.1 | 18 - 19 | 2144 | 98 | ||
| XM_011532003.1 | XP_011530305.1 | 18 - 19 | 2144 | 98 | ||
| XM_011532005.2 | XP_011530307.1 | 18 - 19 | 2144 | 98 | ||
| XM_017008249.1 | XP_016863738.1 | 17 - 18 | 2046 | 98 | ||
| XM_017008250.1 | XP_016863739.1 | 16 - 17 | 2164 | 98 | ||
| XM_017008251.1 | XP_016863740.1 | 17 - 18 | 2046 | 98 | ||
| XM_017008252.1 | XP_016863741.1 | 15 - 16 | 2066 | 98 | ||
| XM_017008253.1 | XP_016863742.1 | 17 - 18 | 1992 | 98 | ||
| XM_017008254.1 | XP_016863743.1 | 10 - 11 | 1102 | 98 | ||
| GenBank mRNA | AB067488.1 | - | 17 - 18 | 2056 | 98 | |
| CR933642.1 | - | 18 - 19 | 2097 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NEK1
Genomic Map