Assay Details
Gene Symbol: ZC3H14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF474376.1 | - | 1 - 2 | 667 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZC3H14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF474376.1 | - | 1 - 2 | 667 | 145 |
Set Membership: |
![]() ![]() ![]() |