Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009418.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009418.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009418.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009418.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009418.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009418.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009418.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009418.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009418.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009418.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009418.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009418.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009418.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009418.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009418.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009418.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170661.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170661.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170661.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170661.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170661.hg.1 |
TC1100009418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100170660.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170660.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170660.hg.1 |
ENST00000527818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170660.hg.1 |
NM_001301851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170660.hg.1 |
TC1100009418.hg.1 |
Clariom D TC |