Assay Details
Gene Symbol: TMEM110-MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL833659.1 | - | 1 - 2 | 4452 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000486659.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000486659.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000486659.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000486659.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | ENST00000486659.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000464769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000464769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000464769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000464769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000477591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000477591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000477591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000477591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000482155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000482155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000485356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000485356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013997.hg.1 | ENST00000485356.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000464769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000464769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000464769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000464769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000477591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000477591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000477591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000477591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000482155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000482155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000485356.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000485356.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013997.hg.1 | ENST00000485356.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000446157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000486659.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000486659.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000486659.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000486659.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | ENST00000486659.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | NM_205853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013995.hg.1 | NM_205853.3 | Refseq |
