Assay Details
| Gene Symbol: MIR99AHG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027790.3 | 2 - 3 | 305 | 114 | ||
| NR_111004.2 | 1 - 2 | 278 | 114 | |||
| NR_111005.2 | 1 - 2 | 278 | 114 | |||
| NR_136541.1 | 2 - 3 | 358 | 114 | |||
| NR_136542.1 | 2 - 3 | 358 | 114 | |||
| GenBank mRNA | AK095614.1 | - | 2 - 3 | 305 | 114 | |
| CN313455.1 | - | 1 - 2 | 278 | 114 | ||
More Information
Set Membership: |
|

MIR99AHG
Genomic Map