Assay Details
| Gene Symbol: ADGRE4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY181245.1 | - | 4 - 5 | 394 | 139 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 121440 | - | - |

ADGRE4P
Genomic Map