Assay Details
Gene Symbol: IMMP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005250630.3 | XP_005250687.1 | 2 - 3 | 141 | 122 | |
XM_017012702.1 | XP_016868191.1 | 2 - 3 | 141 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005250630.3, XM_017012702.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
signal peptide processing
protein processing involved in protein targeting to mitochondrion
superoxide metabolic process
cellular response to DNA damage stimulus
spermatogenesis
brain development
blood circulation
respiratory electron transport chain
ovulation
mitochondrial respiratory chain complex assembly
cerebellum vasculature development
serine-type endopeptidase activity
peptidase activity
ovarian follicle development
signal peptide processing
protein processing involved in protein targeting to mitochondrion
superoxide metabolic process
cellular response to DNA damage stimulus
spermatogenesis
brain development
blood circulation
respiratory electron transport chain
ovulation
mitochondrial respiratory chain complex assembly
cerebellum vasculature development
serine-type endopeptidase activity
peptidase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000437687.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000437687.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000447215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000447215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000447215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000447215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000447215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452753.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452753.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452753.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452753.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000487733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000487733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000487733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000487733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000487733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000492938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000492938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000492938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | ENST00000492938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000331762.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000405709.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000437687.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000437687.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000447215.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000447215.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000447215.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000447215.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000447215.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000450877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452753.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452753.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452753.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452753.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000452895.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000487733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000487733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000487733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000487733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000487733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000489381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000492938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000492938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000492938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | ENST00000492938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_001244606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012255.hg.1 | NM_032549.3 | Refseq |
