Assay Details
Gene Symbol: POPDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017011194.1 | XP_016866683.1 | 1 - 2 | 1591 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017011194.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
