Assay Details
| Gene Symbol: UGT1A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019076.4 | NP_061949.3 | 1 - 2 | 919 | 137 | |
| GenBank mRNA | AF030310.1 | - | 1 - 2 | 856 | 137 | |
| AF462267.1 | - | 1 - 2 | 889 | 137 | ||
| AF462268.1 | - | 1 - 2 | 919 | 137 | ||
| AK313488.1 | - | 1 - 2 | 902 | 137 | ||
| AY435143.1 | - | 1 - 2 | 856 | 137 | ||
| DQ364251.1 | - | 1 - 2 | 511 | 137 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Inactivation of the pure antiestrogen fulvestrant and other synthetic estrogen molecules by UDP-glucuronosyltransferase 1A enzymes expressed in breast tissue.
- Authors
- Chouinard S, Tessier M, Vernouillet G, Gauthier S, Labrie F, Barbier O, Belanger A
- Journal
- Mol Pharmacol - Page No 908 (Wed Mar 01 00:00:00 EST 2006)
- Volume (Year)
- 69 (2006)
- Product Usage
- -

UGT1A8
Genomic Map