Assay Details
| Gene Symbol: RRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301064.1 | NP_001287993.1 | 10 - 11 | 1006 | 87 | |
| NM_018427.4 | NP_060897.3 | 11 - 12 | 1096 | 87 | ||
| XM_005255376.1 | XP_005255433.1 | 9 - 10 | 871 | 87 | ||
| XM_005255377.4 | XP_005255434.1 | 8 - 9 | 820 | 87 | ||
| XM_011522540.2 | XP_011520842.1 | 6 - 7 | 508 | 87 | ||
| XM_011522541.1 | XP_011520843.1 | 10 - 11 | 935 | 87 | ||
| GenBank mRNA | AF227156.1 | - | 11 - 12 | 1062 | 87 | |
| AJ272050.1 | - | 11 - 12 | 1016 | 87 | ||
| AK225662.1 | - | 11 - 12 | 1072 | 87 | ||
| AK295486.1 | - | 8 - 9 | 689 | 87 | ||
| AK300253.1 | - | 10 - 11 | 915 | 87 | ||
| AK302994.1 | - | 8 - 9 | 743 | 87 | ||
| AK303405.1 | - | 7 - 8 | 589 | 87 | ||
| AK304856.1 | - | 11 - 12 | 962 | 87 | ||
| AK314769.1 | - | 11 - 12 | 1010 | 87 | ||
| AK316520.1 | - | 10 - 11 | 920 | 87 | ||
| BC071868.1 | - | 3 - 4 | 331 | 87 | ||
| BC132688.1 | - | 11 - 12 | 1013 | 40146710 | 87 | |
| BC132690.1 | - | 11 - 12 | 1013 | 40146712 | 87 | |
| DA764092.1 | - | 3 - 4 | 299 | 87 | ||
More Information
Set Membership: |
|

RRN3
Genomic Map