Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320589.1 | NP_001307518.1 | 4 - 5 | 681 | 102 | |
| XM_017005412.1 | XP_016860901.1 | 8 - 9 | 1075 | 102 | ||
| GenBank mRNA | BU857820.1 | - | 4 - 5 | 539 | 102 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |

ACYP2
Genomic Map