| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042364.hg.1 |
TC02002643.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042364.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042364.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042366.hg.1 |
TC02002643.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042366.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042366.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042365.hg.1 |
TC02002643.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042365.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042365.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200212215.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200212215.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200212215.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200212215.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200212215.hg.1 |
TC0200015330.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200212213.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200212213.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200212213.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200212213.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200212213.hg.1 |
TC0200015330.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200212214.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200212214.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200212214.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200212214.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200212214.hg.1 |
TC0200015330.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200212216.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200212216.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200212216.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200212216.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200212216.hg.1 |
TC0200015330.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |