Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006161.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006161.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006161.hg.1 |
TC11000517.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006162.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006162.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006162.hg.1 |
TC11000517.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006159.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006159.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006159.hg.1 |
TC11000517.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100154987.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154987.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154987.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154987.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154987.hg.1 |
TC1100007780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100154990.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154990.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154990.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154990.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154990.hg.1 |
TC1100007780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007780.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154991.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154991.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154991.hg.1 |
ENST00000308287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154991.hg.1 |
NM_206893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154991.hg.1 |
TC1100007780.hg.1 |
Clariom D TC |