Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000482428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000482428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000482428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000489578.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000489578.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
ENST00000489578.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020853.hg.1 |
ENST00000331272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020853.hg.1 |
ENST00000389046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020853.hg.1 |
ENST00000470093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020853.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020853.hg.1 |
TC10001609.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006836.hg.1 |
ENST00000426721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006836.hg.1 |
ENST00000426721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006836.hg.1 |
ENST00000426721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006836.hg.1 |
ENST00000426721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006836.hg.1 |
ENST00000426721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006836.hg.1 |
NR_033408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006836.hg.1 |
NR_033408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006836.hg.1 |
NR_033408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006836.hg.1 |
NR_033408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006836.hg.1 |
NR_033408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179645.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179645.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179645.hg.1 |
ENST00000482428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179645.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179645.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179645.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179645.hg.1 |
ENST00000482428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179645.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179645.hg.1 |
TC1000011652.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC2200006836.hg.1 |
ENST00000426721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006836.hg.1 |
ENST00000426721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006836.hg.1 |
ENST00000426721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006836.hg.1 |
ENST00000426721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006836.hg.1 |
ENST00000426721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006836.hg.1 |
NR_033408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006836.hg.1 |
NR_033408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006836.hg.1 |
NR_033408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006836.hg.1 |
NR_033408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006836.hg.1 |
NR_033408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000331272.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000457105.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000470093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000482428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000482428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000482428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000489578.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000489578.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
ENST00000489578.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011652.hg.1 |
NM_022039.3 |
Refseq |