Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005484.hg.1 |
ENST00000322122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005484.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005484.hg.1 |
TC16000373.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005485.hg.1 |
ENST00000322122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005485.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005485.hg.1 |
TC16000373.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000576745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000576745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000576745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600155146.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155146.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600155146.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155146.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600155146.hg.1 |
TC1600007526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000576745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000576745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000576745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007526.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600155145.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155145.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155145.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600155145.hg.1 |
ENST00000322122.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155145.hg.1 |
ENST00000613872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155145.hg.1 |
NM_001008274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600155145.hg.1 |
TC1600007526.hg.1 |
Clariom D TC |