Assay Details
| Gene Symbol: C5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005271889.3 | XP_005271946.1 | 1 - 2 | 893 | 174 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005271889.3 transcripts.
Set Membership: |
|

C5orf24
Genomic Map