Assay Details
Gene Symbol: RP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011515468.2 | XP_011513770.1 | 1 - 2 | 639 | 149 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011515468.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
