Assay Details
Gene Symbol: ZNF724P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045525.1 | 2 - 3 | 262 | 132 | ||
GenBank mRNA | AK301230.1 | - | 2 - 3 | 252 | 132 | |
DC372621.1 | - | 2 - 3 | 262 | 132 | ||
DC391309.1 | - | 2 - 3 | 252 | 132 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001364.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CGH Microarray 4x180K | 8550 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 718326 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 115094 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 892693 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 968644 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 42587 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 72365 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 307373 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 232936 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 38531 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 365801 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P13007373 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P13008430 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P41166112 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P41166151 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8035840 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8035839 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8035841 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8035838 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1900007559.hg.1 | ENST00000593450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007559.hg.1 | ENST00000593450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007559.hg.1 | ENST00000593450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19018993.hg.1 | ENST00000418100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19018993.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19018993.hg.1 | TC19001364.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19018994.hg.1 | ENST00000418100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19018994.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19018994.hg.1 | TC19001364.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010209.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19018996.hg.1 | ENST00000418100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19018996.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19018996.hg.1 | TC19001364.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19018995.hg.1 | ENST00000418100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19018995.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19018995.hg.1 | TC19001364.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007529.hg.1 | ENST00000504759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010209.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900183928.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183928.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900183928.hg.1 | TC1900010209.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900183928.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183928.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900183929.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183929.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183929.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900183929.hg.1 | TC1900010209.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900183929.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183929.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183929.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900183931.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183931.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183931.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183931.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900183931.hg.1 | TC1900010209.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900183931.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183931.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183931.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183931.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900183932.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183932.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183932.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183932.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900183932.hg.1 | TC1900010209.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900183932.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183932.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183932.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183932.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900183934.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183934.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183934.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183934.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900183934.hg.1 | TC1900010209.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900183934.hg.1 | ENST00000418100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183934.hg.1 | ENST00000597037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183934.hg.1 | ENST00000597537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900183934.hg.1 | NR_045525.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007559.hg.1 | ENST00000593450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007559.hg.1 | ENST00000593450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007559.hg.1 | ENST00000593450.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1565748_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1565746_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1565746_at | - | - |
Human Genome U133 Plus 2.0 Array | 1565748_at | - | - |
