Assay Details
Gene Symbol: FAM234B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017019706.1 | XP_016875195.1 | 1 - 2 | 128 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017019706.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000538231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000538231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000538231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000538231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
