Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130963.1 | NP_001124435.1 | 4 - 5 | 588 | 94 | |
| XM_011538056.2 | XP_011536358.1 | 4 - 5 | 1268 | 94 | ||
| XM_011538057.2 | XP_011536359.1 | 5 - 6 | 690 | 94 | ||
| XM_011538059.1 | XP_011536361.1 | 4 - 5 | 442 | 94 | ||
| XM_011538060.2 | XP_011536362.1 | 4 - 5 | 1305 | 94 | ||
| XM_017019077.1 | XP_016874566.1 | 4 - 5 | 1039 | 94 | ||
| XM_017019078.1 | XP_016874567.1 | 4 - 5 | 302 | 94 | ||
| GenBank mRNA | AB006624.2 | - | 4 - 5 | 549 | 94 | |
| AL833310.1 | - | 4 - 5 | 566 | 94 | ||
| AL833311.1 | - | 4 - 5 | 566 | 94 | ||
| BC071597.1 | - | 4 - 5 | 560 | 94 | ||
| DA724501.1 | - | 4 - 5 | 442 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011538059.1, XM_017019078.1 transcripts.
Set Membership: |
|

NEMP1
Genomic Map