Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015257.2 | NP_056072.2 | 3 - 4 | 375 | 114 | |
| XM_011538058.2 | XP_011536360.1 | 3 - 4 | 1052 | 114 | ||
| GenBank mRNA | AK294566.1 | - | 3 - 4 | 347 | 114 | |
| BC117436.1 | - | 3 - 4 | 347 | 40125987 | 114 | |
| BC126300.1 | - | 3 - 4 | 347 | 8992016 | 114 | |
| CR749406.1 | - | 3 - 4 | 347 | 114 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001619.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000553654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000553654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000553654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000553654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021762.hg.1 | ENST00000300128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021762.hg.1 | ENST00000379391.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021762.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021762.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021762.hg.1 | TC12001619.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184849.hg.1 | TC1200010946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000300128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000379391.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000553654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000553654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000553654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000553654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | ENST00000554340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_001130963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_015257.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010946.hg.1 | NM_015257.2 | Refseq |

NEMP1
Genomic Map