Assay Details
| Gene Symbol: RBM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324242.1 | NP_001311171.1 | 4 - 5 | 575 | 131 | |
| NM_001324243.1 | NP_001311172.1 | 4 - 5 | 668 | 131 | ||
| NM_001324244.1 | NP_001311173.1 | 4 - 5 | 575 | 131 | ||
| NR_136728.1 | 4 - 5 | 575 | 131 | |||
| NR_136729.1 | 5 - 6 | 669 | 131 | |||
| NR_136730.1 | 4 - 5 | 575 | 131 | |||
| NR_136731.1 | 4 - 5 | 668 | 131 | |||
| NR_136732.1 | 4 - 5 | 668 | 131 | |||
| XM_011530980.2 | XP_011529282.1 | 4 - 5 | 667 | 131 | ||
| GenBank mRNA | AK023732.1 | - | 4 - 5 | 571 | 131 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000474499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000474499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000474499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000474499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |

RBM41
Genomic Map