Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012816.hg.1 |
ENST00000280557.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012816.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012816.hg.1 |
TC12000976.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012815.hg.1 |
ENST00000280557.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012815.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012815.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012815.hg.1 |
TC12000976.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000537955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000537955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000537955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000537955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000539463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000539463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000539463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000539463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
ENST00000539463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000537955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000537955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000537955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000537955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000539463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000539463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000539463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000539463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
ENST00000539463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009217.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168419.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168419.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168419.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168419.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168419.hg.1 |
ENST00000455982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168419.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168419.hg.1 |
TC1200009217.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200168420.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168420.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168420.hg.1 |
ENST00000280557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168420.hg.1 |
NM_003677.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168420.hg.1 |
TC1200009217.hg.1 |
Clariom D TC |