Assay Details
| Gene Symbol: ZYG11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011541480.2 | XP_011539782.1 | 9 - 10 | 1846 | 145 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000371532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | ENST00000612017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001004339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008334.hg.1 | NM_001307931.1 | Refseq |

ZYG11A
Genomic Map