Assay Details
| Gene Symbol: COX7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001867.2 | NP_001858.1 | 1 - 2 | 164 | 171 | |
| GenBank mRNA | AK026505.1 | - | 1 - 2 | 190 | 171 | |
| BC001005.2 | - | 1 - 2 | 164 | 2821167 | 171 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000427.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC001005.2,NM_001867.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05006019.hg.1 | TC05000427.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006019.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006019.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006019.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006019.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006019.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016964.hg.1 | ENST00000430450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006016.hg.1 | TC05000427.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006016.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006016.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006016.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006016.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006016.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006017.hg.1 | TC05000427.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006017.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006017.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006017.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05006017.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009221.hg.1 | ENST00000532177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008008.hg.1 | NR_037402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008911.hg.1 | ENST00000446556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154758.hg.1 | TC0500008008.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500154760.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154760.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154760.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154760.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154760.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154760.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154760.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154760.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154760.hg.1 | TC0500008008.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008008.hg.1 | NR_037402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016964.hg.1 | ENST00000430450.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008911.hg.1 | ENST00000446556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000247655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000505430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000509578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000510447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000511472.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000513124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | ENST00000515763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | NM_001867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154762.hg.1 | TC0500008008.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100009221.hg.1 | ENST00000532177.1 | Ensembl |

COX7C
Genomic Map