Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007459.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007459.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007459.hg.1 |
TC10000662.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008476.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157616.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157616.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157616.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157616.hg.1 |
ENST00000285949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157616.hg.1 |
ENST00000624358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157616.hg.1 |
NM_183374.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157616.hg.1 |
TC1000008476.hg.1 |
Clariom D TC |