Assay Details
| Gene Symbol: GANC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198141.2 | NP_937784.2 | 15 - 16 | 2059 | 145 | |
| GenBank mRNA | AF525397.1 | - | 11 - 12 | 1312 | 145 | |
| AF545045.1 | - | 15 - 16 | 1753 | 145 | ||
| AF545046.1 | - | 15 - 16 | 1753 | 145 | ||
| AK074037.1 | - | 15 - 16 | 1951 | 145 | ||
| BC059406.1 | - | 15 - 16 | 1931 | 30330796 | 145 | |
| BC093833.1 | - | 15 - 16 | 1776 | 7939678 | 145 | |
| BC112051.1 | - | 15 - 16 | 1776 | 8327519 | 145 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15002792.hg.1
Set Membership: |
|

GANC
Genomic Map