Assay Details
Gene Symbol: CACUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153810.4 | NP_722517.3 | 1 - 2 | 846 | 85 | |
GenBank mRNA | AJ544771.1 | - | 1 - 2 | 630 | 85 | |
AK097728.1 | - | 2 - 3 | 566 | 85 | ||
AY743663.1 | - | 1 - 2 | 846 | 85 | ||
BC038294.2 | - | 1 - 2 | 846 | 4837841 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001694.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK097728.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
positive regulation of cell proliferation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of protein kinase activity
protein binding
protein kinase binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
G1/S transition of mitotic cell cycle
positive regulation of cell proliferation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of protein kinase activity
protein binding
protein kinase binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10022340.hg.1 | ENST00000340214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022340.hg.1 | ENST00000369151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022340.hg.1 | ENST00000369156.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022340.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10022340.hg.1 | TC10001694.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10022343.hg.1 | ENST00000340214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022343.hg.1 | ENST00000369151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022343.hg.1 | ENST00000369156.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022343.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10022343.hg.1 | TC10001694.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10022339.hg.1 | ENST00000340214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022339.hg.1 | ENST00000369151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022339.hg.1 | ENST00000369156.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022339.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10022339.hg.1 | TC10001694.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10022344.hg.1 | ENST00000340214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022344.hg.1 | ENST00000369151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022344.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10022344.hg.1 | TC10001694.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10022342.hg.1 | ENST00000340214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022342.hg.1 | ENST00000369151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022342.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10022342.hg.1 | TC10001694.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10022341.hg.1 | ENST00000340214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022341.hg.1 | ENST00000369151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022341.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10022341.hg.1 | TC10001694.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000181981.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181981.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181981.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181981.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181981.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181981.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181981.hg.1 | TC1000011975.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000181978.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181978.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181978.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181978.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181978.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181978.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181978.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181978.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181978.hg.1 | TC1000011975.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181980.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181980.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181980.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181980.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181980.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181980.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181980.hg.1 | TC1000011975.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000181979.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181979.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181979.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181979.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181979.hg.1 | ENST00000369151.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181979.hg.1 | ENST00000477583.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181979.hg.1 | ENST00000493518.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181979.hg.1 | NM_153810.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181979.hg.1 | TC1000011975.hg.1 | Clariom D TC |
