Assay Details
| Gene Symbol: CACUL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153810.4 | NP_722517.3 | 2 - 3 | 984 | 127 | |
| GenBank mRNA | AJ544771.1 | - | 2 - 3 | 768 | 127 | |
| AK097728.1 | - | 3 - 4 | 704 | 127 | ||
| AY743663.1 | - | 2 - 3 | 984 | 127 | ||
| BC038294.2 | - | 2 - 3 | 984 | 4837841 | 127 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001694.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
positive regulation of cell proliferation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of protein kinase activity
protein binding
protein kinase binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
G1/S transition of mitotic cell cycle
positive regulation of cell proliferation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of protein kinase activity
protein binding
protein kinase binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10022337.hg.1 | ENST00000340214.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022337.hg.1 | ENST00000369151.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022337.hg.1 | ENST00000369156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022337.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022337.hg.1 | TC10001694.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181977.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181977.hg.1 | ENST00000477583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181977.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181977.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181977.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181977.hg.1 | ENST00000477583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181977.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181977.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181977.hg.1 | TC1000011975.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000369151.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000477583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000481360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000489169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000490610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000493518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | ENST00000544392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011975.hg.1 | NM_153810.4 | Refseq |

CACUL1
Genomic Map