Assay Details
Gene Symbol: SP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003111.4 | NP_003102.1 | 2 - 3 | 689 | 103 | |
GenBank mRNA | AK304199.1 | - | 2 - 3 | 261 | 103 | |
BC126414.1 | - | 2 - 3 | 211 | 8992130 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
