Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003530.hg.1 |
ENST00000245304.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003530.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003530.hg.1 |
TC13000341.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003532.hg.1 |
ENST00000245304.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003532.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003532.hg.1 |
TC13000341.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003535.hg.1 |
ENST00000245304.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003535.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003535.hg.1 |
TC13000341.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003534.hg.1 |
ENST00000245304.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003534.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003534.hg.1 |
TC13000341.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007780.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007780.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152353.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152353.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152353.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152353.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152353.hg.1 |
TC1300007780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300152355.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152355.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152355.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152355.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152355.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152355.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152355.hg.1 |
TC1300007780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300152358.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152358.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152358.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152358.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152358.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152358.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152358.hg.1 |
TC1300007780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300152359.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152359.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152359.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152359.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152359.hg.1 |
TC1300007780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300152357.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152357.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152357.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152357.hg.1 |
ENST00000245304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152357.hg.1 |
ENST00000476869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152357.hg.1 |
NM_021033.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152357.hg.1 |
TC1300007780.hg.1 |
Clariom D TC |