Assay Details
| Gene Symbol: GPR137C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099652.1 | NP_001093122.1 | 3 - 4 | 711 | 163 | |
| GenBank mRNA | AK123953.1 | - | 3 - 4 | 607 | 163 | |
| AK293812.1 | - | 1 - 2 | 160 | 163 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000307.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003233.hg.1 | ENST00000321662.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003233.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003233.hg.1 | TC14000307.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007168.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | ENST00000321662.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | ENST00000542169.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | ENST00000555369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | ENST00000555622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | NM_001099652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400149721.hg.1 | TC1400007168.hg.1 | Clariom D TC |

GPR137C
Genomic Map