Assay Details
| Gene Symbol: ATP8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105529.1 | NP_001098999.1 | 14 - 15 | 1527 | 145 | |
| XM_011513618.1 | XP_011511920.1 | 14 - 15 | 1522 | 145 | ||
| XM_017007645.1 | XP_016863134.1 | 14 - 15 | 1556 | 145 | ||
| XM_017007647.1 | XP_016863136.1 | 13 - 14 | 1427 | 145 | ||
| GenBank mRNA | AB209687.1 | - | 14 - 15 | 1380 | 145 | |
| BC109317.1 | - | 14 - 15 | 1289 | 40036536 | 145 | |
| BC109318.1 | - | 14 - 15 | 1295 | 40036538 | 145 | |
More Information
Set Membership: |
|

ATP8A1
Genomic Map